3-(Difluoromethyl)-2-fluoropyridine 95%

Lieferant: Apollo Scientific

PC502292-10G PC502292-1G PC502292-5G
APOSPC502292-10GEA 561 EUR
APOSPC502292-10G APOSPC502292-1G APOSPC502292-5G
3-(Difluoromethyl)-2-fluoropyridine 95%
3-(Difluoromethyl)-2-fluoropyridine
Formel: C₆H₄F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD22199298
CAS-Nummer: 1374659-42-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR