5-(Difluoromethyl)-2-fluoropyridine 97%

Lieferant: Apollo Scientific

PC502322-10G PC502322-1G PC502322-250MG
APOSPC502322-10GEA 0 EUR
APOSPC502322-10G APOSPC502322-1G APOSPC502322-250MG
5-(Difluoromethyl)-2-fluoropyridine 97%
5-(Difluoromethyl)-2-fluoropyridine
Formel: C₆H₄F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD22199299
CAS-Nummer: 1374659-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR