Ethyl 3-bromo-2,4,5-trifluorobenzoylacetate
Lieferant: Apollo Scientific
PC502395-1G
APOSPC502395-1GEA
1180
EUR
APOSPC502395-1G
Ethyl 3-bromo-2,4,5-trifluorobenzoylacetate
Ethyl 3-bromo-2,4,5-trifluorobenzoylacetate
Formel:
C₁₁H₈BrF₃O₃ Molecular Weight: 325,079 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12965122 CAS-Nummer: 104222-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...