4,5-Difluoro-2-hydroxybenzoic acid 95+%

Lieferant: Apollo Scientific

PC502404-1G PC502404-250MG
APOSPC502404-1GEA 226 EUR
APOSPC502404-1G APOSPC502404-250MG
4,5-Difluoro-2-hydroxybenzoic acid 95+%
4,5-Difluoro-2-hydroxybenzoic acid
Formel: C₇H₄F₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13185554
CAS-Nummer: 205533-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR