3,6-Difluoro-2-iodoaniline 95%

Lieferant: Apollo Scientific
Achtung

PC502426-1G PC502426-250MG
APOSPC502426-1GEA 1090 EUR
APOSPC502426-1G APOSPC502426-250MG
3,6-Difluoro-2-iodoaniline 95%
3,6-Difluoro-2-iodoaniline
Formel: C₆H₄F₂IN
Molecular Weight: 255.004 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28789179
CAS-Nummer: 1582732-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR