3,6-Difluoro-2-nitrophenol 97%
Lieferant: Apollo Scientific
PC502443-1G
APOSPC502443-1GEA
186
EUR
APOSPC502443-1G
3,6-Difluoro-2-nitrophenol 97%
3,6-Difluoro-2-nitrophenol
Formel:
C₆H₃F₂NO₃ Molecular Weight: 175,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22042170 CAS-Nummer: 139548-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...