2,6-Difluoro-3-(trifluoromethoxy)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC502453-250MG
PC502453-1G
PC502453-5G
APOSPC502453-250MGEA
174
EUR
APOSPC502453-250MG
APOSPC502453-1G
APOSPC502453-5G
2,6-Difluoro-3-(trifluoromethoxy)benzoic acid 95%
2,6-Difluoro-3-(trifluoromethoxy)benzoic acid
Formel:
C₈H₃F₅O₃ |
MDL:
MFCD31561155 CAS-Nummer: 2244077-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...