2,6-Difluoro-3-(trifluoromethoxy)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC502453-250MG PC502453-1G PC502453-5G
APOSPC502453-250MGEA 174 EUR
APOSPC502453-250MG APOSPC502453-1G APOSPC502453-5G
2,6-Difluoro-3-(trifluoromethoxy)benzoic acid 95%
2,6-Difluoro-3-(trifluoromethoxy)benzoic acid
Formel: C₈H₃F₅O₃
MDL: MFCD31561155
CAS-Nummer: 2244077-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR