3-Fluoro-4-(trifluoromethoxy)styrene 95%

Lieferant: Apollo Scientific
Achtung

PC502476-1G PC502476-250MG
APOSPC502476-1GEA 371 EUR
APOSPC502476-1G APOSPC502476-250MG
3-Fluoro-4-(trifluoromethoxy)styrene 95%
3-Fluoro-4-(trifluoromethoxy)styrene
Formel: C₉H₆F₄O
Lagertemperatur: Kühlschrank
MDL: MFCD30184382
CAS-Nummer: 2168352-66-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR