2,3-Difluoro-6-(trifluoromethoxy)benzyl alcohol 95%
Lieferant: Apollo Scientific
Achtung
|
PC502493-1G
APOSPC502493-1GEA
609
EUR
APOSPC502493-1G
2,3-Difluoro-6-(trifluoromethoxy)benzyl alcohol 95%
2,3-Difluoro-6-(trifluoromethoxy)benzyl alcohol
Formel:
C₈H₅F₅O₂ Molecular Weight: 228.12 g/mol |
MDL:
MFCD31561164 CAS-Nummer: 2244081-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...