2,3-Difluoro-6-(trifluoromethoxy)benzyl bromide 95%
Lieferant: Apollo Scientific
Gefahr
|
PC502496-1G
APOSPC502496-1GEA
609
EUR
APOSPC502496-1G
2,3-Difluoro-6-(trifluoromethoxy)benzyl bromide 95%
2,3-Difluoro-6-(trifluoromethoxy)benzyl bromide
Formel:
C₈H₄BrF₅O |
MDL:
MFCD31561165 CAS-Nummer: 2244083-21-2 UN: 3265 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...