6-Bromo-4-fluorobenzo[b]thiophene

Lieferant: Apollo Scientific
Achtung

PC502503-1G
APOSPC502503-1GEA 928 EUR
APOSPC502503-1G
6-Bromo-4-fluorobenzo[b]thiophene
6-Bromo-4-fluorobenzo[b]thiophene
Formel: C₈H₄BrFS
Molecular Weight: 231.08 g/mol
MDL: MFCD23705320
CAS-Nummer: 826995-63-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR