2,6-Difluoro-4-(trifluoromethoxy)benzonitrile 95%

Lieferant: Apollo Scientific
Achtung

PC502516-1G PC502516-250MG PC502516-5G
APOSPC502516-1GEA 522 EUR
APOSPC502516-1G APOSPC502516-250MG APOSPC502516-5G
2,6-Difluoro-4-(trifluoromethoxy)benzonitrile 95%
2,6-Difluoro-4-(trifluoromethoxy)benzonitrile
Formel: C₈H₂F₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD28791009
CAS-Nummer: 1806305-45-2
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR