2,6-Difluoro-4-(trifluoromethoxy)benzonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC502516-1G
PC502516-250MG
PC502516-5G
APOSPC502516-1GEA
522
EUR
APOSPC502516-1G
APOSPC502516-250MG
APOSPC502516-5G
2,6-Difluoro-4-(trifluoromethoxy)benzonitrile 95%
2,6-Difluoro-4-(trifluoromethoxy)benzonitrile
Formel:
C₈H₂F₅NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD28791009 CAS-Nummer: 1806305-45-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...