2,3-Difluoro-4-(trifluoromethoxy)benzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC502521-1G PC502521-250MG
APOSPC502521-1GEA 567 EUR
APOSPC502521-1G APOSPC502521-250MG
2,3-Difluoro-4-(trifluoromethoxy)benzaldehyde 95%
2,3-Difluoro-4-(trifluoromethoxy)benzaldehyde
Formel: C₈H₃F₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31561154
CAS-Nummer: 2244086-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR