5-Bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

PC502549-1G
APOSPC502549-1GEA 518 EUR
APOSPC502549-1G
5-Bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
5-Bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
Formel: C₁₀H₄BrF₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD23115255
CAS-Nummer: 826995-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR