6-Bromo-7-fluorobenzo[b]thiophene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC502571-1G
APOSPC502571-1GEA
1030
EUR
APOSPC502571-1G
6-Bromo-7-fluorobenzo[b]thiophene 95%
6-Bromo-7-fluorobenzo[b]thiophene
Formel:
C₈H₄BrFS Molecular Weight: 231,08 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD23705326 CAS-Nummer: 1427437-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...