Methyl 5-fluoro-2-methoxybenzoate

Lieferant: Apollo Scientific

PC502593-1G
APOSPC502593-1GEA 49.3 EUR
APOSPC502593-1G
Methyl 5-fluoro-2-methoxybenzoate
Methyl 5-fluoro-2-methoxybenzoate
Formel: C₉H₉FO₃
Molecular Weight: 184,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203734
CAS-Nummer: 151793-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR