2-Fluor-4-nitrobenzonitril

Lieferant: Apollo Scientific

PC502594-25G
APOSPC502594-25GEA 87 EUR
APOSPC502594-25G
2-Fluor-4-nitrobenzonitril
2-Fluor-4-nitrobenzonitril
Formel: C₇H₃FN₂O₂
Molecular Weight: 166,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038715
CAS-Nummer: 34667-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR