Methyl-2-fluor-5-nitrobenzoat 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-fluoro-5-nitrobenzenecarboxylate, Methyl 2-fluoro-5-nitrobenzoate
PC502595-25G
APOSPC502595-25GEA
63.8
EUR
APOSPC502595-25G
Methyl-2-fluor-5-nitrobenzoat 98%
Methyl-2-fluoro-5-nitrobenzenecarboxylate
Formel:
C₈H₆FNO₄ Molecular Weight: 199.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203689 CAS-Nummer: 2965-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...