3-Iod-4-(trifluormethoxy)benzoesäure 95+%

Lieferant: Apollo Scientific
Achtung

PC502616-250MG
APOSPC502616-250MGEA 82.8 EUR
APOSPC502616-250MG
3-Iod-4-(trifluormethoxy)benzoesäure 95+%
3-Iodo-4-(trifluoromethoxy)benzoic acid
Formel: C₈H₄F₃IO₃
Molecular Weight: 332,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08459287
CAS-Nummer: 1110709-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR