2-Nitro-5-(trifluoromethyl)benzaldehyde 97%

Lieferant: Apollo Scientific

PC502647-250MG
APOSPC502647-250MGEA 97.3 EUR
APOSPC502647-250MG
2-Nitro-5-(trifluoromethyl)benzaldehyde 97%
2-Nitro-5-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13185768
CAS-Nummer: 1176723-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR