Methyl 3-nitro-4-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific

PC502651-5G
APOSPC502651-5GEA 56.7 EUR
APOSPC502651-5G
Methyl 3-nitro-4-(trifluoromethyl)benzoate
Methyl 3-nitro-4-(trifluoromethyl)benzoate
Formel: C₉H₆F₃NO₄
Molecular Weight: 249.143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264510
CAS-Nummer: 126541-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR