4-Nitro-3-(trifluormethyl)benzonitril

Lieferant: Apollo Scientific

PC502658-25G PC502658-5G
APOSPC502658-25GEA 215 EUR
APOSPC502658-25G APOSPC502658-5G
4-Nitro-3-(trifluormethyl)benzonitril
4-Nitro-3-(trifluormethyl)benzonitril
Formel: C₈H₃F₃N₂O₂
Molecular Weight: 216.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664270
CAS-Nummer: 320-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR