2-Nitro-5-(trifluoromethoxy)benzonitrile

Lieferant: Apollo Scientific

PC502663-1G PC502663-5G
APOSPC502663-1GEA 168 EUR
APOSPC502663-1G APOSPC502663-5G
2-Nitro-5-(trifluoromethoxy)benzonitrile
2-Nitro-5-(trifluoromethoxy)benzonitrile
Formel: C₈H₃F₃N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09835205
CAS-Nummer: 1003708-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR