2-Bromo-6-(trifluoromethoxy)benzyl bromide ≥95%

Lieferant: Apollo Scientific

PC502679-250MG PC502679-1G PC502679-5G
APOSPC502679-250MGEA 143 EUR
APOSPC502679-250MG APOSPC502679-1G APOSPC502679-5G
2-Bromo-6-(trifluoromethoxy)benzyl bromide ≥95%
2-Bromo-6-(trifluoromethoxy)benzyl bromide
Formel: C₈H₅Br₂F₃O
MDL: MFCD31561241
CAS-Nummer: 1253189-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR