2-Bromo-6-(trifluoromethoxy)benzaldehyde
Lieferant: Apollo Scientific
PC502684-1G
PC502684-5G
PC502684-10G
APOSPC502684-1GEA
60.9
EUR
APOSPC502684-1G
APOSPC502684-5G
APOSPC502684-10G
2-Bromo-6-(trifluoromethoxy)benzaldehyde
2-Bromo-6-(trifluoromethoxy)benzaldehyde
Formel:
C₈H₄BrF₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11856022 CAS-Nummer: 1114809-17-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...