Methyl 3-bromo-5-(trifluoromethoxy)benzoate

Lieferant: Apollo Scientific

PC502691-1G PC502691-5G
APOSPC502691-1GEA 61.3 EUR
APOSPC502691-1G APOSPC502691-5G
Methyl 3-bromo-5-(trifluoromethoxy)benzoate
Methyl 3-bromo-5-(trifluoromethoxy)benzoate
Formel: C₉H₆BrF₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09999511
CAS-Nummer: 1306763-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR