Methyl 5-bromo-2-(trifluoromethoxy)benzoate
Lieferant: Apollo Scientific
PC502692-1G
APOSPC502692-1GEA
294
EUR
APOSPC502692-1G
Methyl 5-bromo-2-(trifluoromethoxy)benzoate
Methyl 5-bromo-2-(trifluoromethoxy)benzoate
Formel:
C₉H₆BrF₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203799 CAS-Nummer: 773874-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...