5-Bromo-2-(trifluoromethoxy)benzyl bromide 95%

Lieferant: Apollo Scientific

PC502694-1G PC502694-5G
APOSPC502694-1GEA 143 EUR
APOSPC502694-1G APOSPC502694-5G
5-Bromo-2-(trifluoromethoxy)benzyl bromide 95%
5-Bromo-2-(trifluoromethoxy)benzyl bromide
Formel: C₈H₅Br₂F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD03701032
CAS-Nummer: 685126-87-8
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR