4-Bromo-2-(trifluoromethoxy)benzoic acid

Lieferant: Apollo Scientific

PC502695-25G
APOSPC502695-25GEA 85.7 EUR
APOSPC502695-25G
4-Bromo-2-(trifluoromethoxy)benzoic acid
4-Bromo-2-(trifluoromethoxy)benzoic acid
Formel: C₈H₄BrF₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12546979
CAS-Nummer: 509142-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR