Methyl 2-chloro-6-(trifluoromethoxy)benzoate 95%

Lieferant: Apollo Scientific
Achtung

PC502714-1G PC502714-250MG
APOSPC502714-1GEA 0 EUR
APOSPC502714-1G APOSPC502714-250MG
Methyl 2-chloro-6-(trifluoromethoxy)benzoate 95%
Methyl 2-chloro-6-(trifluoromethoxy)benzoate
Formel: C₉H₆ClF₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18399023
CAS-Nummer: 1261453-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR