2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-nitrophenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(2-fluoro-4-nitro-phenoxy)-5-(trifluoromethyl)benzene

PC502728-1G
APOSPC502728-1GEA 1180 EUR
APOSPC502728-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-nitrophenyl ether
Formel: C₁₃H₅F₆NO₃
Molecular Weight: 337,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629955
CAS-Nummer: 2244085-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR