2-Chloro-4-fluoro-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Chloro-4-fluoro-5-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502729-1G
APOSPC502729-1GEA 1180 EUR
APOSPC502729-1G
2-Chloro-4-fluoro-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Chloro-4-fluoro-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄ClF₆NO₃
Molecular Weight: 371,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629959
CAS-Nummer: 2244085-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR