2-Bromo-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Bromo-6-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502731-1G
APOSPC502731-1GEA 1180 EUR
APOSPC502731-1G
2-Bromo-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅BrF₅NO₃
Molecular Weight: 398,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629966
CAS-Nummer: 2244085-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR