4-Bromo-2-chloro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Bromo-2-chloro-6-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502732-1G
APOSPC502732-1GEA 1180 EUR
APOSPC502732-1G
4-Bromo-2-chloro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-2-chloro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄BrClF₅NO₃
Molecular Weight: 432,525 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629969
CAS-Nummer: 2244088-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR