2,6-Difluoro-4-(trifluoromethyl)phenyl 4-methoxy-2-nitrophenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(4-methoxy-2-nitro-phenoxy)-5-(trifluoromethyl)benzene

PC502734-1G
APOSPC502734-1GEA 1180 EUR
APOSPC502734-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-methoxy-2-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-methoxy-2-nitrophenyl ether
Formel: C₁₄H₈F₅NO₄
Molecular Weight: 349,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629975
CAS-Nummer: 2244085-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR