2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluoro-4-nitrophenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-2-(3-fluoro-4-nitro-phenoxy)-5-(trifluoromethyl)benzene

PC502737-1G
APOSPC502737-1GEA 1180 EUR
APOSPC502737-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluoro-4-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-fluoro-4-nitrophenyl ether
Formel: C₁₃H₅F₆NO₃
Molecular Weight: 337,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629984
CAS-Nummer: 2244083-25-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR