2,6-Difluoro-4-(trifluoromethyl)phenyl 2-methyl-3-nitrophenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
1,3-Difluoro-2-(2-methyl-3-nitro-phenoxy)-5-(trifluoromethyl)benzene
PC502740-1G
APOSPC502740-1GEA
1180
EUR
APOSPC502740-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-methyl-3-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-methyl-3-nitrophenyl ether
Formel:
C₁₄H₈F₅NO₃ Molecular Weight: 333,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31629993 CAS-Nummer: 2244083-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...