2-Bromo-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Bromo-5-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502742-1G
APOSPC502742-1GEA 1180 EUR
APOSPC502742-1G
2-Bromo-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅BrF₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31629999
CAS-Nummer: 2244086-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR