4-Bromo-3-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Bromo-3-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502743-1G
APOSPC502743-1GEA 1180 EUR
APOSPC502743-1G
4-Bromo-3-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-3-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅BrF₅NO₃
Molecular Weight: 398,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630002
CAS-Nummer: 2244085-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR