2,6-Difluoro-4-(trifluoromethyl)phenyl 3-methyl-2-nitrophenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
1,3-Difluoro-2-(3-methyl-2-nitro-phenoxy)-5-(trifluoromethyl)benzene
PC502745-1G
APOSPC502745-1GEA
1180
EUR
APOSPC502745-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-methyl-2-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 3-methyl-2-nitrophenyl ether
Formel:
C₁₄H₈F₅NO₃ Molecular Weight: 333,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630008 CAS-Nummer: 2244088-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...