2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-5-methyl-2-nitrophenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
1-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-4-fluoro-5-methyl-2-nitrobenzene
PC502750-1G
APOSPC502750-1GEA
1180
EUR
APOSPC502750-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-5-methyl-2-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-5-methyl-2-nitrophenyl ether
Formel:
C₁₄H₇F₆NO₃ Molecular Weight: 351,201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630023 CAS-Nummer: 2244086-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...