2,3-Difluoro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 3-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-1,2-difluoro-4-nitrobenzene

PC502752-1G
APOSPC502752-1GEA 1180 EUR
APOSPC502752-1G
2,3-Difluoro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2,3-Difluoro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄F₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31630029
CAS-Nummer: 2244088-18-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR