5-Bromo-3-fluoro-2-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(5-Bromo-3-fluoro-2-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502753-1G
APOSPC502753-1GEA 1180 EUR
APOSPC502753-1G
5-Bromo-3-fluoro-2-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-3-fluoro-2-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄BrF₆NO₃
Molecular Weight: 416,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630032
CAS-Nummer: 2244083-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR