2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methyl-5-nitrophenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
1-[2,6-Difluoro-4-(trifluoromethyl)phenoxy]-2-fluoro-4-methyl-5-nitrobenzene
PC502754-1G
APOSPC502754-1GEA
1180
EUR
APOSPC502754-1G
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methyl-5-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methyl-5-nitrophenyl ether
Formel:
C₁₄H₇F₆NO₃ Molecular Weight: 351,201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630035 CAS-Nummer: 2244085-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...