2-Bromo-3-fluoro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

PC502756-1G
APOSPC502756-1GEA 1180 EUR
APOSPC502756-1G
2-Bromo-3-fluoro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-3-fluoro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄BrF₆NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31630041
CAS-Nummer: 2244088-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR