4-Bromo-2-fluoro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Bromo-2-fluoro-6-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502757-1G
APOSPC502757-1GEA 1180 EUR
APOSPC502757-1G
4-Bromo-2-fluoro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-2-fluoro-6-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₄BrF₆NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31630044
CAS-Nummer: 2244088-48-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR