2-Bromo-5-fluoro-4-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-(2-Bromo-5-fluoro-4-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene
PC502758-1G
APOSPC502758-1GEA
1180
EUR
APOSPC502758-1G
2-Bromo-5-fluoro-4-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-5-fluoro-4-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel:
C₁₃H₄BrF₆NO₃ Molecular Weight: 416,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630047 CAS-Nummer: 2244087-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...