3-Bromo-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Bromo-5-nitro-phenoxy)-1,3-difluoro-5-(trifluoromethyl)benzene

PC502759-1G
APOSPC502759-1GEA 1180 EUR
APOSPC502759-1G
3-Bromo-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Bromo-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formel: C₁₃H₅BrF₅NO₃
Molecular Weight: 398,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630050
CAS-Nummer: 2244085-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR